Anti-MAD1L1 Mouse Monoclonal Antibody [clone: 9B10]

GTX25783
89361-734EA 481.14 CAD
89361-734
Anti-MAD1L1 Mouse Monoclonal Antibody [clone: 9B10]
Mouse monoclonal antibody [9B10] to MAD1L1

Recommended Starting Dilutions:WB: Use at 1:200 - 1:1,000ELISA: Use at 1:5,000 - 1:20,000IP: Use at 1:100ICC/IF: Use at a concentration of 10-20 ?g/mL. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MAD1L1
Clonality: Monoclonal
Clone: 9B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR