Anti-MAGEA1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX16034
89351-204EA 618.87 CAD
89351-204
Anti-MAGEA1 Rabbit Polyclonal Antibody
The MAGE1 gene is a member of the melanoma antigen-encoding gene family.

IHC-P: Working dilution: Ready-to-use for 10 minutes at RT. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM citrate buffer, pH 6.0 for 10 minutes followed by cooling at RT for 20 minutes. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MAGEA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR