Anti-MAGEA1 Mouse Monoclonal Antibody [clone: E22-11B2-E9]

Supplier: GeneTex

GTX23211
89351-928EA 640.8 CAD
89351-928
Anti-MAGEA1 Mouse Monoclonal Antibody [clone: E22-11B2-E9]
Antibodies
Mouse monoclonal [E22 - 11B2 - E9] to MAGE 1

Western Blotting: use at a concentration of 1 - 2?g / ml for 2hrs at RT. Not tested in other applications. Optimal dilutions / concentrations should be determined by the end user. By Western blot, this antibody detects a band of 42 - 46 kDa, which corresponds to the predicted molecular weight of MAGE 1.

Type: Primary
Antigen: MAGEA1
Clonality: Monoclonal
Clone: E22-11B2-E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR