Anti-CBP Rabbit Polyclonal Antibody

Supplier: Genetex
GTX10489
89350-780EA 466.68 CAD
89350-780
Anti-CBP Rabbit Polyclonal Antibody
Cyclic AMP-responsive enhancer binding protein (CREB) binding protein (CBP) and p300 are closely related transcriptional coactivators that have been shown to directly interact with many different DNA-binding transcription factors including nuclear hormone receptors, CREB (cyclic AMP-responsive enhancer binding protein), c-Fos, c-Jun/v-Jun, c-Myb/v-Myb, TFIIB and MyoD.Both CBP and p300 have been shown to display histone acetyltransferase (HAT) activity, capable of acetylating all four core histone particles in nucleosomes.As a result of HAT activity, it has been suggested CBP and p300 may play a direct role in activating chromatin for transcription.Single point mutations in CBP have been proposed as causative factors in the developmental abnormalities of Rubinstein-Taybi syndrome (RTS).Although both CBP and p300 appear to function similarly, the inability of p300 to rescue CBP malfunction iRTS suggests intrinsic functional differences between CBP and p300.

Type: Primary
Antigen: CBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR