Anti-CBP Rabbit Polyclonal Antibody

GTX77886
89366-380EA 518.15 CAD
89366-380
Anti-CBP Rabbit Polyclonal Antibody
CBP/p300-interacting transactivator 1 (CITED1, melanocyte-specific protein 1 ) is a nuclear protein that shares two highly conserved domains, CR1 and CR2. The CR2 domain is significantly acidic and activates transcription in yeast cells.

Type: Primary
Antigen: CBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR