Anti-MDC1 Rabbit Polyclonal Antibody

GTX11170
89350-802EA 606.37 CAD
89350-802
Anti-MDC1 Rabbit Polyclonal Antibody
The MDC1 protein contains a forkhead homology-associated (FHA) domain and 2 BRCA1 C-terminal motifs (BRCTs) and is required for the intra-S-phase DNA damage checkpoint.

Type: Primary
Antigen: MDC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR