Anti-MED28 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX85388
89353-924EA 920.76 CAD
89353-924
Anti-MED28 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to MED28

Type: Primary
Antigen: MED28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR