Anti-MAP2K1 + MAP2K2 Rabbit Monoclonal Antibody

GTX61116
89363-832EA 585.93 CAD
89363-832
Anti-MAP2K1 + MAP2K2 Rabbit Monoclonal Antibody
MEK1 and MEK2 (MAPK kinase 1/2, or ERK kinase 1/2) are mitogen-activated protein kinases that stimulate MAP kinase activity, playing a role in both cell growth and differentiation . MEK itself is activated via phosphorylation at serines 217/218 and 221/222 by upstream activator kinases, including c-raf, mos and MEK kinase . MEK1 and MEK2 are activated by a wide variety of growth factors and cytokines, and also by membrane depolarization and calcium influx .

Type: Primary
Antigen: MAP2K1 + MAP2K2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR