Anti-Mitochondrial P110 Mouse Monoclonal Antibody [clone: 2G2]

Supplier: GeneTex
GTX10435
89350-296EA 672.9 CAD
89350-296
Anti-Mitochondrial P110 Mouse Monoclonal Antibody [clone: 2G2]
Antibodies
p110 has an approximate isoelectric point of 6.5 and co-partitions with HSP-60 proteins during isolation of mitochondria from HeLa cells. Close examination of the staining pattern in HeLa and Fanconiis anemia cells reveal differences in the morphology and organization of mitochondria in these two cell types. In HeLa cells, mitochondria appear as individual tubular compartments of variable length and are closely associated with vimentin filaments, particularly at the periphery of the nucleus. In Fanconiis anemia cells, mitochondria have a filamentous shape and form an interconnected cytoplasmic reticulum running in parallel with both vimentin filaments and microtubules.

Type: Primary
Antigen: Mitochondrial P110
Clonality: Monoclonal
Clone: [2G2]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR