Anti-MLCK Rabbit Monoclonal Antibody

Supplier: Genetex
GTX61685
89364-128EA 585.93 CAD
89364-128
Anti-MLCK Rabbit Monoclonal Antibody
Myosin light chain kinase (MLCK) is a serine/threonine kinase that belongs to CaM Kinases (Ca++/Calmodulin-dependent protein kinases) family. MLCK is involved in the regulation of myosin activity during smooth muscle contraction. At increased level of Ca2+, rapid binding of Ca2+ to Calmodulin (CaM) occurs, triggering MLCK activation. Once activated, MLCK phosphorylates myosin light chain (MLC) and leading to cross-bridges cycling and generation of the contractile force . MLCK has been implicated in the regulation of endothelial and vascular permeability and in signaling pathway which leads to fibroblast apoptosis .

Type: Primary
Antigen: MLCK
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR