Anti-MMP11 Mouse Monoclonal Antibody

Supplier: Genetex
GTX24102
89352-112EA 650.55 CAD
89352-112
Anti-MMP11 Mouse Monoclonal Antibody
Matrix metalloproteinase (MMPs) are a family of closely related zinc containing proteolytic enzymes, capable of degrading connective tissue components. MMP 11 is localized on the long arm of chromosome 22. It was originally identified on the basis that it is expressed specifically in stromal cells surrounding invasive breast carcinomas. MMP 11 is expression is found more specifically in malignant tumors than in benign ones.

Type: Primary
Antigen: MMP11
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR