Anti-MMP3 Mouse Monoclonal Antibody [clone: SL-1 ID3]

Supplier: GeneTex
GTX23204
89351-926EA 565.28 CAD
89351-926
Anti-MMP3 Mouse Monoclonal Antibody [clone: SL-1 ID3]
Antibodies
Mouse monoclonal [SL - 1 ID3] to MMP3

Not suitable for IHC. For WB: Use at a dilution of 1:400. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: MMP3
Clonality: Monoclonal
Clone: SL-1 ID3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR