Anti-MMP8 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX113780
89357-730EA 881.1 CAD
89357-730
Anti-MMP8 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to MMP8 (matrix metallopeptidase 8 (neutrophil collagenase))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MMP8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR