Anti-MMP9 Rabbit Polyclonal Antibody

GTX74518
89365-460EA 512.33 CAD
89365-460
Anti-MMP9 Rabbit Polyclonal Antibody
Recognizes the carboxyterminal region of matrix metalloproteinase 9 (MMP-9). React with other MMP family members. In quiescent cells, MMP-9 may be expressed at low levels. It has reported that some tumour cell lines constitutively produce MMP-9.

Type: Primary
Antigen: MMP9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR