Anti-IgA Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX29170
89352-596EA 356.34 CAD
89352-596
Anti-IgA Rabbit Polyclonal Antibody
Antibodies
IgA is the predominant immunoglobulin class in body secretions, such as saliva, tears, bronchial secretions, nasal mucosal secretions, prostatic fluid, vaginal secretions, and mucous secretions of the small intestines. It may serve both to defend against local infection and to prevent access of foreign antigens to the general immunologic system. This secondary antibody specifically recognises Mouse IgA and does not cross react with immunoglobulin subclasses.

Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR