Anti-IgA Goat Polyclonal Antibody

Supplier: GeneTex
GTX77223
89353-550EA 264.14 CAD
89353-550
Anti-IgA Goat Polyclonal Antibody
Antibodies
The affinity purified antibodies react only with the IgA in serum and secretions by immunodiffusion and IEP techniques. Antibodies are suitable for Elisa and Blotting Applications. Goats were immunized with highly purified mouse serum IgA in Freunds adjuvant. The resulting antiserum was solid-phase absorbed to make it IgA alpha chain specific. The anti IgA alpha antibodies were immunoaffinity purified off an IgA-containing immunoabsorbant and conjugated with horseradish peroxidase.

Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR