Anti-IgG Rat Monoclonal Antibody [clone: LO-MG3-7] (Biotin)

Supplier: GeneTex

GTX11657
89358-660EA 351.62 CAD
89358-660
Anti-IgG Rat Monoclonal Antibody [clone: LO-MG3-7] (Biotin)
Antibodies
Immunoglobulin G (IgG), is one of the most abundant proteins in human serum with normal levels between 8-17 mg/ml in adult blood. IgG is important for our defence against microorganisms and the molecules are produced by B lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions; to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. The variability of the IgG pool is generated by somatic recombination and the number of specificities in an individual at a given time point is estimated to be 1011 variants.

Recommended Starting Dilutions:ELISA: Use at a concentration of 0.5 - 1.0 µg/mL.Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy chain
Host: Rat
Isotype: IgM
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR