Goat Anti-Mouse IgA/IgG/IgM Polyclonal Antibody

GTX26007
89352-150EA 294.83 CAD
89352-150
Goat Anti-Mouse IgA/IgG/IgM Polyclonal Antibody
This secondary antibody specifically recognizes Mouse IgG + IgM + IgA. It does not react with any other isotypes.

Type: Secondary
Antigen: IgA/IgG/IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR