Anti-IgG Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX27000
89352-218EA 469.22 CAD
89352-218
Anti-IgG Rabbit Polyclonal Antibody
Antibodies
Can be used in immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR