Anti-MPG Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX101916
89349-352EA 881.1 CAD
89349-352
Anti-MPG Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to MPG (N-methylpurine-DNA glycosylase)

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MPG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR