Anti-MPO Mouse Monoclonal Antibody [clone: 18B7]

Supplier: GeneTex
GTX82941
89353-874EA 1483.78 CAD
89353-874
Anti-MPO Mouse Monoclonal Antibody [clone: 18B7]
Antibodies
Mouse monoclonal antibody [18B7] to Myeloperoxidase

For ELISA: Use at an assay dependent dilution. Can be used as a capture antibody in an ELISA pair with GTX82940 (detection). For WB: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: MPO
Clonality: Monoclonal
Clone: 18B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR