Anti-MRP14/S100-A9 Rabbit Monoclonal Antibody

GTX62293
89364-382EA 556.26 CAD
89364-382
Anti-MRP14/S100-A9 Rabbit Monoclonal Antibody
S100-A9 (MRP14) is a member of the S100 family proteins containing two EF-hand calcium-binding motifs . S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation . S100-A9 forms a heterodimer, Calprotectin, with S100-A8 in a calcium-dependent manner. Calprotectin is a major calcuim- and zinc-binding protein in the cytosol of neutrophils, monocytes, and keratinocytes . S100-A9 may function in the inhibition of casein kinase and altered expression of this protein is associated with the disease cystic fibrosis .

Type: Primary
Antigen: MRP14/S100-A9
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR