Anti-ABCC11 Rabbit Monoclonal Antibody

GTX62287
89364-380EA 520.01 CAD
89364-380
Anti-ABCC11 Rabbit Monoclonal Antibody
Rabbit monoclonal antibody [EPR3554] to MRP8 / S100A8

Is unsuitable for Flow Cyt or ICC/IF. IHC-P: 1/500 - 1/1000. Perform heat mediated antigen retrieval via the pressure cooker method before commencing with IHC staining protocol. IP: 1/30. WB: 1/1000.

Type: Primary
Antigen: ABCC11
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR