Anti-MSH gamma 3 Rabbit Polyclonal Antibody

GTX11139
89356-710EA 563 CAD
89356-710
Anti-MSH gamma 3 Rabbit Polyclonal Antibody
MSH (Melanocyte Stimulating Hormone) increases the pigmentation of skin by increasing melanin production in melanocytes.

Type: Primary
Antigen: MSH gamma 3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chimpanzee
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR