Anti-MUC1 Chicken Polyclonal Antibody

GTX82408
89368-078EA 642.96 CAD
89368-078
Anti-MUC1 Chicken Polyclonal Antibody
Chicken polyclonal to Mucin-1

IHC: 1:200WB: 1:500*Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: MUC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR