Anti-MUC16 Mouse Monoclonal Antibody [clone: OV185:1]

Supplier: GeneTex
GTX20697
89351-534EA 700.32 CAD
89351-534
Anti-MUC16 Mouse Monoclonal Antibody [clone: OV185:1]
Antibodies
Several studies have shown that CA125 is useful tumor marker for ovarian epithelial malignancies. However, CA 125 has also been described in other neoplasms such as seminal vesicle and anaplastic lymphomas.

For IHC-P: Use at a dilution of 1:25-1:50 for 30 min at RT in an ABC method. Requires high temperature antigen unmasking with 10mM citrate buffer, pH 6.0 prior to immunostaining. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: MUC16
Clonality: Monoclonal
Clone: OV185:1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR