Anti-MUC16 Goat Polyclonal Antibody

GTX88166
89366-958EA 402.96 CAD
89366-958
Anti-MUC16 Goat Polyclonal Antibody
Thought to provide a protective, lubricating barrier against particles and infectious agents at mucosal surfaces.

Type: Primary
Antigen: MUC16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR