Anti-MUC3 Mouse Monoclonal Antibody

GTX17744
89351-364EA 521.39 CAD
89351-364
Anti-MUC3 Mouse Monoclonal Antibody
Mucin 3 (MUC3) is a glycoprotein. distributed in colon and rectum, and to a lesser extent in breast, lung and salivary gland tissues. This antibody shows no cross-reaction with MUC1 and MUC2.

Type: Primary
Antigen: MUC3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR