Anti-TRIM54 Rabbit Polyclonal Antibody

GTX108065
89355-272EA 544.95 CAD
89355-272
Anti-TRIM54 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to MURF3 (tripartite motif-containing 54)

Suggested starting dilutions are as follows: IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: TRIM54
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR