Anti-MSC Goat Polyclonal Antibody

Supplier: GeneTex

GTX21330
89361-008EA 673.38 CAD
89361-008
Anti-MSC Goat Polyclonal Antibody
Antibodies
Gene Ontology terms - transcription co-repressor activity; transcription factor activity; regulation of transcription, DNA-dependent; transcription from Pol II promoter; nucleus. Principal Names - MSC; ABF1; MYOR; ABF-1; musculin (activated B-cell factor-1); musculin; activated B-cell factor-1; activated B-cell factor 1, homolog of mouse musculin. Official Gene Symbol - MSC GenBank Accession Number ~ NP_005089 LocusLink ID - 9242 (human).

Western Blot: use at 1.0 - 3.0 ?g/ml. Not tested in other applications. Optimal dilutions / concentrations should be determined by the end user. By Western blot, a 20 - 25 kDa band is seen in MOLT 4 lysate. Predicted MW of 24 kDa according to NP_005089.

Type: Primary
Antigen: MSC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR