Anti-MB Goat Polyclonal Antibody

Supplier: Genetex
GTX19209
89360-582EA 544.8 CAD
89360-582
Anti-MB Goat Polyclonal Antibody
Myoglobin is a small heme containing protein (153 amino acid residues, molecular weight (w/o heme) 17053 Da and theoretical pI=7.29) responsible for the oxygen deposition in muscle tissues. Only one form of myoglobin is expressed in cardiac and skeletal muscles. Myoglobin is known as a marker of myocardial damage and it has been used for more than three decades. Nowadays it still is very commonly used in clinical practice as an early marker of AMI. It appears in patient's blood 1 to 3 hours after onset of the symptoms, reaching peak level within 8 to 12 hours. Myoglobin is not so cardiac specific as cTnI or cTnT. Because of high myoglobin concentration in skeletal muscle tissue, even minor skeletal muscle injury results in the significant increase of myoglobin concentration in blood. Thus myoglobin is used together with cTnI or cTnT in clinical practise for better specificity in AMI diagnosis.

Type: Primary
Antigen: MB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR