Anti-MHC1 and MHC2 Mouse Monoclonal Antibody

GTX17811
89351-396EA 551.26 CAD
89351-396
Anti-MHC1 and MHC2 Mouse Monoclonal Antibody
SM-MHC is a cytoplasmic structural protein, which is a major component of the contractile apparatus in smooth muscle cells. Expression of smooth muscle myosin is developmently regulated, appearing early in smooth muscle development, and is specific for smooth muscle development. Two isoforms of smooth muscle myosin heavy chain have been identified, designated MHC-1 and MHC-2. This antibody reacts with MHC-1 (205kDa) and MHC-2 (200kDa) isoform, and it stains the intact myoepithelial cell layers present in benign and in situ malignant breast lesions.

Type: Primary
Antigen: MHC1 and MHC2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR