Anti-Myosin Skeletal + SMOOTH Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX11085
89356-478EA 998.26 CAD
89356-478
Anti-Myosin Skeletal + SMOOTH Rabbit Polyclonal Antibody
Antibodies
Myosin is a 500 kD protein known to interact with actin in muscle and in non-muscle cells. It contains two identical heavy chains (200 kD each) and four light chains (15-20 kD each). Myosin molecules consist of two major regions; the tail (rod) and head. These molecules aggregate into filaments through the tail region and interact with actin and ATP through the head region.

Type: Primary
Antigen: Myosin Skeletal + SMOOTH
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Cow (Bovine)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR