Anti-KAT8 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX104587
89350-394EA 264.05 CAD
89350-394
Anti-KAT8 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to MYST1 (MYST histone acetyltransferase 1)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: KAT8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR