Anti-NAT1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX113619
89357-658EA 708.98 CAD
89357-658
Anti-NAT1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to NAT1 (N-acetyltransferase 1 (arylamine N-acetyltransferase))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR