Anti-NCK Mouse Monoclonal Antibody [clone: NC-20 (same as 20B-1H9)]

GTX23215
89351-930EA 595.42 CAD
89351-930
Anti-NCK Mouse Monoclonal Antibody [clone: NC-20 (same as 20B-1H9)]
Mouse monoclonal [NC - 20 ( same as 20B - 1H9 )] to Nck

For IP (Native and Denatured, Use Protein G): Use at a dilution of 1:400 of protein lysate. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: NCK
Clonality: Monoclonal
Clone: NC-20 (same as 20B-1H9)
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR