Anti-NDC80 Rabbit Monoclonal Antibody

GTX63082
89364-626EA 556.26 CAD
89364-626
Anti-NDC80 Rabbit Monoclonal Antibody
Ndc80 is one of several proteins involved in spindle checkpoint signaling. This surveillance mechanism assures correct segregation of chromosomes during cell division by detecting unaligned chromosomes and causing prometaphase arrest until the proper bipolar attachment of chromosomes is achieved.

Type: Primary
Antigen: NDC80
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR