Anti-NDUFS1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX105270
89354-180EA 903.1 CAD
89354-180
Anti-NDUFS1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to NDUFS1 (NADH dehydrogenase (ubiquinone) Fe-S protein 1, 75kDa (NADH-coenzyme Q reductase))

Suggested starting dilutions are as follows: WB: 1:500-1:10000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NDUFS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR