Anti-NEDD8 Mouse Monoclonal Antibody [clone: ND23]

Supplier: GeneTex
GTX30772
89362-992EA 748.01 CAD
89362-992
Anti-NEDD8 Mouse Monoclonal Antibody [clone: ND23]
Antibodies
Mouse monoclonal [ND23] to Nedd8

For ELISA: Use at an assay dependent dilution. For ICC: Use at an assay dependent dilution. For WB: Use at a concentration of 0.15-0.3 ?g/ml (determined by immunoblotting of recombinant GST-Nedd8). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user

Type: Primary
Antigen: NEDD8
Clonality: Monoclonal
Clone: ND23
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR