Anti-Neuro D1 Rabbit Monoclonal Antibody

GTX62615
89364-530EA 556.26 CAD
89364-530
Anti-Neuro D1 Rabbit Monoclonal Antibody
NeuroD1 belongs to the NeuroD family of basic helix-loop-helix (bHLH) transcription factors. NeuroD1 forms heterodimers with other bHLH proteins and activates transcription of genes that contain a specific DNA sequence known as the E-box. It regulates expression of insulin, and mutations in NeuroD1 result in type II diabetes mellitus .

Type: Primary
Antigen: Neuro D1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR