Anti-NRP1 Rabbit Monoclonal Antibody [clone: EPR3113]

GTX62190
89364-344EA 520.01 CAD
89364-344
Anti-NRP1 Rabbit Monoclonal Antibody [clone: EPR3113]
Rabbit monoclonal antibody [EPR3113] to Neuropilin-1

Recommended Starting Dilutions: For WB: Use at a dilution of 1:1000 For IHC: Use at a dilution of 1:100 For FACS: Use at a dilution of 1:100 For IP: Use at a dilution of 1:50 Optimal working dilution for a specific application should be determined by the investigator.

Type: Primary
Antigen: NRP1
Clonality: Monoclonal
Clone: EPR3113
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Monkey, Rat, Marmoset
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR