Anti-NF1 Rabbit Polyclonal Antibody

GTX77638
89368-016EA 638.09 CAD
89368-016
Anti-NF1 Rabbit Polyclonal Antibody
GTX77638 is specific for human Neurofibromin 1 (NF1), the protein product of the NF1 tumour suppressor gene.

Type: Primary
Antigen: NF1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR