Anti-NFKB P50 Rabbit Polyclonal Antibody

GTX73125
89353-144EA 446.07 CAD
89353-144
Anti-NFKB P50 Rabbit Polyclonal Antibody
NF kappa B is a heterodimer consisting of a 50 kD DNA binding subunit and a 65 kD transactivating subunit (RelA). p50 subunit of NFkB has an isoform and both the proteins are derived from larger precursor proteins of 105 and 100 kD respectively.

Type: Primary
Antigen: NFKB P50
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR