Anti-RELA Rabbit Monoclonal Antibody

GTX61804
89364-198EA 585.93 CAD
89364-198
Anti-RELA Rabbit Monoclonal Antibody
Nuclear factor B (NF-B) encompasses an important family of inducible transcriptional activators that regulate a wide variety of cellular and viral genes . The family members include p50, p52, p65 (RelA), c-Rel, and RelB. . The p65 subunit, similar to two others in the B family, RelB and c-Rel, contains two transactivation domains in the C-terminal region of the protein . Association with inhibitory proteins of the IκB family retains NF-κB in the cytoplasm. Degradation of IB proteins exposes the nuclear localization sequence (NLS), leading to nuclear translocation and subsequent binding of NF-B to DNA . p65 phosphorylation events occur in the cytoplasm or in the nucleus and are stimuli-specific and cell-type specific, on serine 276, 529, and 536 by various kinases, such as PKA, casein kinase II, IKK beta, and RSK1. Phosphorylation on these sites has been linked to regulation of NF-κB activation .

Type: Primary
Antigen: RELA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR