Anti-NEFM Chicken Polyclonal Antibody

Supplier: GeneTex
GTX85461
89353-934EA 996.89 CAD
89353-934
Anti-NEFM Chicken Polyclonal Antibody
Antibodies
Chicken polyclonal antibody to Neurofilament Medium Chain (NFM) (Neuron cell marker)

Recommended starting dilutions: For IHC/IF: Use at 1:1000 - 1:2500, for WB: Use at 1:2500 - 1:5000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NEFM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR