Anti-NGF Rabbit Monoclonal Antibody [clone: EP1320Y]

Supplier: GeneTex
GTX61496
89364-006EA 583.61 CAD
89364-006
Anti-NGF Rabbit Monoclonal Antibody [clone: EP1320Y]
Antibodies
Rabbit monoclonal antibody [EP1320Y] to NGF beta

Recommended Starting Dilutions: For WB: Use at a dilution of 1:500 For IHC: Use at a dilution of 1:50 100 For ICC: Use at a dilution of 1:100 -250 Optimal working dilution for a specific application should be determined by the investigator.

Type: Primary
Antigen: NGF
Clonality: Monoclonal
Clone: EP1320Y
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR