Anti-Nitrotyrosine Goat Polyclonal Antibody

Supplier: GeneTex
GTX40117
89363-098EA 1006.79 CAD
89363-098
Anti-Nitrotyrosine Goat Polyclonal Antibody
Antibodies
Nitric oxide (NO) is a product of the enzymatic conversion of arginine to citrulline by nitric oxide synthase. NO reacts rapidly with superoxide to form peroxynitrite. At physiological pH and in the presence of transition metals, peroxynitrite undergoes heterolytic cleavage to form hydroxyl anion and nitronium ion, the latter of which nitrates protein tyrosine residues. Thus, the presence of nitrotyrosine on proteins can be used as a marker for peroxynitrite formation in vivo. The presence of nitrotyrosine has been detected in various inflammatory processes including atherosclerotic placques.

Type: Primary
Antigen: Nitrotyrosine
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR