Anti-NKX3A Rabbit Polyclonal Antibody

GTX12044
89359-126EA 503.02 CAD
89359-126
Anti-NKX3A Rabbit Polyclonal Antibody
NKX3A is a member of the NK3 homeobox family and has several isoforms. It is a putative transcription factor and may act as a transcriptional repressor that exhibits prostate and testis specific expression. It may play a role in proliferation of glandular epithelium and in formation of ducts in the prostate. NKX3A is regulated by androgens and estrogen in the prostate cancer cell line LNCaP.

Type: Primary
Antigen: NKX3A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR