Anti-NM23 Mouse Monoclonal Antibody

GTX75469
89365-816EA 440.75 CAD
89365-816
Anti-NM23 Mouse Monoclonal Antibody
Recognises human NM23-HI protein, also known as nucleoside diphosphate kinase A (NDPK-A). There is minor cross-reactivity with the homologous protein NDPK-B (nm23-H2). NM23 expression has been reported to correlate with improved prognosis in mammary carcinoma. The protein is implicated as a metastasis suppressor, and expression is linked with less aggressive forms of this disease.

Type: Primary
Antigen: NM23
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR