Anti-GRIN2A Rabbit Monoclonal Antibody

GTX63442
89364-644EA 587.6 CAD
89364-644
Anti-GRIN2A Rabbit Monoclonal Antibody
NMDAR2A belongs to the glutamante-gated ion channel family (1). This receptor has been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning (2). NMDAR2A forms heteromeric channel of a zeta subunit (GRIN1), a epsilon subunit (GRIN2A, GRIN2B, GRIN2C or GRIN2D) and a third subunit (GRIN3A or GRIN3B). Activation of NMDAR2A requires binding of agonist to both types of subunits (1).

Type: Primary
Antigen: NMDAR2A
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR